missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN18 Monoclonal Antibody (OTI2D3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA505242S
This item is not returnable.
View return policy
Description
ZSCAN18 Monoclonal antibody specifically detects ZSCAN18 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| ZSCAN18 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9H161 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 60529 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI2D3 | |
| Unconjugated | |
| ALX4 | |
| EG232875; RGD1560671; similar to hypothetical protein FLJ12895; zinc finger and SCAN domain containing 18; zinc finger and SCAN domain-containing protein 18; Zinc finger protein 447; ZNF447; ZSCAN18 | |
| Full length human recombinant protein of human ALX4 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction