missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZSCAN2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100769
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ZSCAN2.
May be involved in transcriptional regulation during the post-meiotic stages of spermatogenesis.
Specifications
| ZSCAN2 | |
| Polyclonal | |
| Unconjugated | |
| ZSCAN2 | |
| Zfp29; Zfp-29; zinc finger and SCAN domain containing 2; zinc finger and SCAN domain-containing protein 2; Zinc finger protein 29; zinc finger protein 29 homolog; Zinc finger protein 854; ZNF854; Zscan2 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 22691, 54993 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q07230, Q7Z7L9 | |
| ZSCAN2 | |
| A synthesized peptide derived from human ZSCAN2(Accession Q7Z7L9), corresponding to amino acid residues I322-S372. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction