missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN21 Monoclonal Antibody (OTI2F5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA506146S
This item is not returnable.
View return policy
Description
ZSCAN21 Monoclonal antibody specifically detects ZSCAN21 in Human samples. It is validated for Western BlotSpecifications
| ZSCAN21 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P17813 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 2022 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI2F5 | |
| Unconjugated | |
| ENG | |
| AI326272; CTfin51; hypothetical protein LOC527103; NY-REN-21; renal carcinoma antigen NY-REN-21; RU49; Transcription factor RU49; Zfp38; zfp-38; zinc finger and SCAN domain containing 21; zinc finger and SCAN domain-containing protein 21; zinc finger and SCAN domain-containing protein 21; zinc finger protein 3; zinc finger protein with KRAB and SCAN domains 1; zinc finger proliferation 1; zinc finger protein 38; zinc finger protein 38 (KOX 25); zinc finger protein 38 homolog; zinc finger protein NY-REN-21 antigen; zinc finger with KRAB and SCAN domains 1; Zipro1; ZKSCAN1; ZNF38; ZSCAN21 | |
| Full length human recombinant protein of human ENG produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction