missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSCAN21/ZFP38 Antibody (OTI2E5), Alexa Fluor™ 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-74956AF594
This item is not returnable.
View return policy
Description
ZSCAN21/ZFP38 Monoclonal antibody specifically detects ZSCAN21/ZFP38 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence
Specifications
| ZSCAN21/ZFP38 | |
| Monoclonal | |
| Alexa Fluor 594 | |
| DKFZp434L134, KOX25, NY-REN-21, Renal carcinoma antigen NY-REN-21, Zfp-38, ZFP38, zinc finger and SCAN domain containing 21, zinc finger and SCAN domain-containing protein 21, zinc finger protein 38, zinc finger protein 38 (KOX 25), Zinc finger protein 38 homolog, zinc finger protein NY-REN-21 antigen, Zipro1, ZNF38DKFZp686H10254 | |
| Full length recombinant protein of human ZSCAN21/ZFP38 (NP_666019) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat, Canine, Monkey | |
| Purified |
| Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunofluorescence | |
| OTI2E5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 7589 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction