missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZSCAN4C Polyclonal Antibody
GREENER_CHOICE

Product Code. 15983055
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15983055 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15983055 Supplier Invitrogen™ Supplier No. PA576536

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

Embryonic stem (ES) cell-specific transcription factor required to regulate ES cell pluripotency. Binds telomeres and plays a key role in genomic stability in ES cells by regulating telomere elongation. Acts as an activator of spontaneous telomere sister chromatid exchange (T-SCE) and telomere elongation in undifferentiated ES cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZSCAN4C
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Zscan4c
Gene Accession No. Q80VJ6
Gene Alias Gm397; XM_142517; zinc finger and SCAN domain containing 4C; zinc finger and SCAN domain containing protein 4C; Zscan4c; Zscan4d
Gene Symbols Zscan4c
Host Species Rabbit
Immunogen Synthetic peptide of mouse ZSCAN4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245109
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.