missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZSWIM2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-21055
This item is not returnable.
View return policy
Description
ZSWIM2 Polyclonal specifically detects ZSWIM2 in Human samples. It is validated for Western Blot.
Specifications
| ZSWIM2 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| centromere/kinetochore protein zw10 homolog, HZW10, KNTC1AP, MGC149821, zeste white 10 homolog, ZW10 (Drosophila) homolog, centromere/kinetochore protein, ZW10 homolog, centromere/kinetochore protein, ZW10, kinetochore associated, homolog (Drosophila) | |
| Rabbit | |
| 73 kDa | |
| 0.1 mL | |
| Zinc Finger | |
| 151112 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
| Western Blot | |
| 1mg/mL | |
| Western Blot 1:500-1:3000 | |
| Q8NEG5 | |
| ZSWIM2 | |
| Recombinant protein encompassing a sequence within the center region of human ZSWIM2. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
For Research Use Only