Learn More
Invitrogen™ ZSWIM2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA531253
Description
Recommended positive controls: NT2D1, IMR32, U87-MG, MCF-7. Predicted reactivity: Human (98%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
E3 ubiquitin-protein ligase involved in the regulation of Fas-, DR3- and DR4-mediated apoptosis. Functions in conjunction with the UBE2D1, UBE2D3 and UBE2E1 E2 ubiquitin-conjugating enzymes.
Specifications
| ZSWIM2 | |
| Polyclonal | |
| Unconjugated | |
| ZSWIM2 | |
| E3 ubiquitin-protein ligase ZSWIM2; MEKK1-related protein X; MEX; RING-type E3 ubiquitin transferase ZSWIM2; Zinc finger SWIM domain-containing protein 2; zinc finger SWIM-type containing 2; zinc finger, SWIM domain containing 2; zinc finger, SWIM-type containing 2; ZSWIM2; ZZ-type zinc finger-containing protein 2; ZZZ2 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 151112 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8NEG5 | |
| ZSWIM2 | |
| Recombinant protein encompassing a sequence within the center region of human ZSWIM2. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.