missing translation for 'onlineSavingsMsg'
Learn More

ZW10 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody

€ 280.00 - € 624.00

Specifications

Antigen ZW10
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18613736
View Documents
Novus Biologicals
NBP2-38644-25ul
25 μL
€ 280.00
25µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18138297
View Documents
Novus Biologicals
NBP2-38644
0.1 mL
€ 624.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

ZW10 Polyclonal specifically detects ZW10 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications

Specifications

ZW10
Polyclonal
Rabbit
Mitotic Regulators
PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide
E3 ubiquitin-protein ligase ZSWIM2, EC 6.3.2.-, MEKK1-related protein X, MEX, MGC33890, Zinc finger SWIM domain-containing protein 2, zinc finger, SWIM domain containing 2, zinc finger, SWIM-type containing 2, ZZZ2ZZ-type zinc finger-containing protein 2
ZW10
IgG
Affinity Purified
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
RUO
Human
O43264
9183
This antibody was developed against a recombinant protein corresponding to amino acids: TSTVPLAEMLGDMIWEDLSECLIKNCLVYSIPTNSSKLQQYEEIIQSTEEFENALKEMRFLKGDTTDLLKYARNINSHFANKKCQDVIVAARNLMTSE
Primary
Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.