missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZWINT Monoclonal Antibody (GT276)

Product Code. 15398260
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15398260 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15398260 Supplier Invitrogen™ Supplier No. MA517284

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Recommended positive controls: 293T, A431, HeLa, A375. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that is clearly involved in kinetochore function although an exact role is not known. It interacts with ZW10, another kinetochore protein, possibly regulating the association between ZW10 and kinetochores. The encoded protein localizes to prophase kinetochores before ZW10 does and it remains detectable on the kinetochore until late anaphase. It has a uniform distribution in the cytoplasm of interphase cells. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZWINT
Applications Western Blot
Classification Monoclonal
Clone GT276
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ZWINT
Gene Accession No. O95229
Gene Alias 2010007E07Rik; 2600001N01Rik; D10Ertd749e; human ZW10 interacting protein-1; HZwint-1; KNTC2AP; neuroprotective protein 8; scoilin; Sip30; SNAP25 interacting protein 30; SNAP25 interacting protein of 30 kDa; SNAP25-interacting protein 30; ZW10 interacting kinetochore protein; ZW10 interacting protein-1; ZW10 interactor; ZW10 interactor, kinetochore protein; ZW10-interacting protein 1; ZWINT; ZWINT1; Zwint-1
Gene Symbols ZWINT
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human ZWINT. The exact sequence is proprietary.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11130
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.