missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ZWINT Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA592903

Product Code. 16588291

  • € 530.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: MEAAETEAEA AALEVLAEVA GILEPVGLQE EAELPAKILV EFVVDSQKKD KLLCSQLQVA DFLQNILAQE DTAKGLDPLA SEDTSRQKAI AAKEQWKELK ATYREHVEAI KIGLTKALTQ MEEAQRKRTQ LREAFEQLQA KKQMAMEKRR AVQNQWQLQQ EKHLQHLAEV SAEVRERKTG TQQELDRVFQ KLGNLKQQAE QERDKLQRYQ TFLQLLYTLQ GKLLFPEAEA EAENLPDDKP QQPTRPQEQS TGDTMGRDPG VSFKAVGLQP AGDVNLP; Positive Samples: 293T, SW620, HepG2, Jurkat, Mouse brain; Cellular Location: Chromosome, Nucleus, centromere, kinetochore.

This gene encodes a protein that is clearly involved in kinetochore function although an exact role is not known. It interacts with ZW10, another kinetochore protein, possibly regulating the association between ZW10 and kinetochores. The encoded protein localizes to prophase kinetochores before ZW10 does and it remains detectable on the kinetochore until late anaphase. It has a uniform distribution in the cytoplasm of interphase cells. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZWINT
Polyclonal
Unconjugated
ZWINT
2010007E07Rik; 2600001N01Rik; D10Ertd749e; human ZW10 interacting protein-1; HZwint-1; KNTC2AP; neuroprotective protein 8; scoilin; Sip30; SNAP25 interacting protein 30; SNAP25 interacting protein of 30 kDa; SNAP25-interacting protein 30; ZW10 interacting kinetochore protein; ZW10 interacting protein-1; ZW10 interactor; ZW10 interactor, kinetochore protein; ZW10-interacting protein 1; ZWINT; ZWINT1; Zwint-1
Rabbit
Affinity Chromatography
RUO
11130, 257644, 52696
-20°C, Avoid Freeze/Thaw Cycles
Liquid
ELISA, Western Blot, Immunocytochemistry
2.01 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O95229, Q8VIL3, Q9CQU5
ZWINT
Recombinant fusion protein containing a sequence corresponding to amino acids 1-277 of human ZWINT (NP_0089882).
100 μL
Primary
Human, Mouse, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.