missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZXDA Antibody, DyLight 550, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-04129R
This item is not returnable.
View return policy
Description
ZXDA Polyclonal antibody specifically detects ZXDA in Human samples. It is validated for Western Blot
Specifications
| ZXDA | |
| Polyclonal | |
| Western Blot | |
| zinc finger X-linked protein ZXDA, zinc finger, X-linked, duplicated A, ZNF896 | |
| The immunogen recognized by this antibody maps to this antibody and Zinc Finger, X-linked, Duplicated B using the numbering given in entry NP_009087.1 and NP_009088.1 (GeneID 7789 and 158586). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 550 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 7789 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction