missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ZXDC Antibody (PCRP-ZXDC-2B5), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24184MFV450
This item is not returnable.
View return policy
Description
ZXDC Monoclonal antibody specifically detects ZXDC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ZXDC | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| dJ337O18.5, SWIM domain containing 1, zinc finger, SWIM-type containing 1 | |
| Recombinant full-length human ZXDC protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| PCRP-ZXDC-2B5 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 79364 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction