missing translation for 'onlineSavingsMsg'
Learn More

ZXDC Antibody (PCRP-ZXDC-2B5), Novus Biologicals™

Product Code. 18674345 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µg
20µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18674345 100 μg 100µg
18691334 20 μg 20µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18674345 Supplier Novus Biologicals Supplier No. NBP323621100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

ZXDC Monoclonal antibody specifically detects ZXDC in Human samples. It is validated for Flow Cytometry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen ZXDC
Applications Flow Cytometry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone PCRP-ZXDC-2B5
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Flow Cytometry : 1-2 μg/million cells, Immunocytochemistry/ Immunofluorescence : 1-2 μg/mL, Immunohistochemistry-Paraffin : 1-2 μg/mL
Formulation 10 mM PBS with 0.05% BSA
Gene Alias dJ337O18.5, SWIM domain containing 1, zinc finger, SWIM-type containing 1
Host Species Mouse
Immunogen Recombinant full-length human ZXDC protein
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 79364
Target Species Human
Content And Storage Store at 4°C.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.