missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zyxin Antibody (CL2502) [Alexa Fluor™ 594], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36768AF594
This item is not returnable.
View return policy
Description
Zyxin Monoclonal antibody specifically detects Zyxin in Human samples. It is validated for Immunohistochemistry,Immunohistochemistry (Paraffin),Immunocytochemistry/ Immunofluorescence
Specifications
| Zyxin | |
| Monoclonal | |
| Alexa Fluor 594 | |
| CTCL tumor antigen se14-3, CTCL-associated antigen se14-3, cutaneous T-cell lymphoma associated antigen se14-3, Cutaneous T-cell lymphoma-associated antigen se14-3, KIAA1125, MGC31836, predicted protein of HQ2893, PRKCBP1, PRO2893, protein kinase C-binding protein 1, Rack7, RACK7protein kinase C binding protein 1, zinc finger MYND domain containing protein 8, Zinc finger MYND domain-containing protein 8, zinc finger, MYND-type containing 8 | |
| This antibody was developed using a recombinant protein derived from Q15942, with the exact immunogen sequence remaining proprietary. | |
| 0.1 mL | |
| Signal Transduction | |
| 7791 | |
| Store at 4⪚C in the dark. | |
| IgG2b |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| CL2502 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction