missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Zyxin Antibody, Novus Biologicals™
Goat Polyclonal Antibody
Brand: Novus Biologicals NBP1-46135
This item is not returnable.
View return policy
Description
Zyxin Polyclonal specifically detects Zyxin in Human samples. It is validated for Western Blot, Peptide ELISA.
Specifications
| Zyxin | |
| Polyclonal | |
| Unconjugated | |
| Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
| CTCL tumor antigen se14-3, CTCL-associated antigen se14-3, cutaneous T-cell lymphoma associated antigen se14-3, Cutaneous T-cell lymphoma-associated antigen se14-3, KIAA1125, MGC31836, predicted protein of HQ2893, PRKCBP1, PRO2893, protein kinase C-binding protein 1, Rack7, RACK7protein kinase C binding protein 1, zinc finger MYND domain containing protein 8, Zinc finger MYND domain-containing protein 8, zinc finger, MYND-type containing 8 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Reported variants represent identical protein: NP_003452.1, NP_001010972.1 | |
| Store at -20C. Avoid freeze-thaw cycles. |
| Western Blot, ELISA | |
| 0.5 mg/mL | |
| Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:4000 | |
| NP_003452.1 | |
| ZYX | |
| Peptide with sequence C-QQREKPRVQEKQH corresponding to internal region according to NP_003452.1. | |
| 0.1 mg | |
| Signal Transduction | |
| 7791 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction