missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZZZ3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA564266
This item is not returnable.
View return policy
Description
Immunogen sequence: LTKAGIPVPG RTPNLYIYSK KSSTSRRQHP LNKHLFKPST FMTSHEPPVY MDEDDDRSCF HSHMNTAVED ASDDESIPIM Y Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 90%.
Component of the ATAC complex, a complex with histone acetyltransferase activity on histones H3 and H4.
Specifications
| ZZZ3 | |
| Polyclonal | |
| Unconjugated | |
| ZZZ3 | |
| 3110065C23Rik; 6430567E01Rik; AA408566; AI481057; ATAC component 1 homolog; ATAC1; AW556059; DKFZP564I052; R75514; RGD1565468; zinc finger ZZ-type containing 3; zinc finger, ZZ domain containing 3; zinc finger, ZZ-type containing 3; ZZ-type zinc finger-containing protein 3; Zzz3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 26009 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8IYH5 | |
| ZZZ3 | |
| Recombinant protein corresponding to Human ZZZ3. Recombinant protein control fragment (Product #RP-103877). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction