Learn More
Thermo Scientific™ Autosampler Sample Tube and Standards Racks
Customize your autosampler with a range of autosampler sample tube and standards racks to suit your requirements.
Brand: Thermo Scientific BRE0033171
Additional Details : Weight : 0.01000kg
Description
Customize your autosampler with a range of autosampler sample tube and standards racks to suit your requirements.
Match Your CETAC Sample Tube Rack to Your Laboratory’s Needs- 21-position sample tube rack
- 24-position sample tube rack
- 40-position sample tube rack
- 60-position sample tube rack
- 90-position sample tube rack
- Extended Rack for CETAC ASX-520 Autosampler (for use with Thermo Scientific™ iCAP™ Q Series ICP-MS systems)
Specifications
Autosampler Sample Tube Racks | |
60-Position Sample Tube Rack | |
Thermo Scientific iSC-65 Autosampler, Teledyne CETAC Autosamplers, ESI Autosamplers |