Learn More
Thermo Scientific™ Autosampler Sample Tube and Standards Racks
Customize your autosampler with a range of autosampler sample tube and standards racks to suit your requirements.
€ 66.43
Product Code | Brand | For Use With (Equipment) | Type | Description | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | For Use With (Equipment) | Type | Description | Price | Quantity & Availability | |||
17881145
|
Thermo Scientific
BRE0033171 |
Thermo Scientific iSC-65 Autosampler, Teledyne CETAC Autosamplers, ESI Autosamplers | Autosampler Sample Tube Racks | 60-Position Sample Tube Rack |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Customize your autosampler with a range of autosampler sample tube and standards racks to suit your requirements.
Choose sample tube and standards racks that meet your laboratory needs
- 21-Position Sample Tube Rack for Teledyne CETAC and ESI Autosamplers
- 24-Position Sample Tube Rack for Teledyne CETAC and ESI Autosamplers
- 40-Position Sample Tube Rack for Teledyne CETAC and ESI Autosamplers
- 60-Position Sample Tube Rack for Teledyne CETAC and ESI Autosamplers
- 90-Position Sample Tube Rack for Teledyne CETAC and ESI Autosamplers
- Standards Rack for 7x250 mL Bottles for ESI SC-4 DX Autosampler
- Standards Rack for 9x125 mL Bottles for ESI SC-2 DX Autosampler
- Standards Rack for 9x125 mL Bottles for ESI SC-4/14 DX Autosampler
- Standards Rack for 4x125 mL Bottles for ESI SC-8 DX Autosampler
- Standards Rack for 10x50 mL Vials for ESI SC-2/4/8/14 DX Autosamplers