Learn More
Huenersdorf™ CLASSIC-FLEX™ Assortment Boxes
These assortment boxes are manufactured of extra dimensionally stable transparent polystyrene for individual fitting with insert boxes.
Brand: Huenersdorf™ 613300
Additional Details : Weight : 0.50000kg
Description
- Transparent lid and bottom for optimum display of contents
- With stable hinges and closures
- Ideal for packaging purposes
- Food-safe and temperature resistant from −20° to +70°C
- Without compartment subdivisions for individual fitting with insert boxes
Specifications
Transparent | |
Storage | |
-20° to +70°C |
Rectangular | |
Insert boxes | |
225 x 335 x 73 mm |