missing translation for 'onlineSavingsMsg'
Learn More

Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs

Features easy-to-grip head for manual assembly. Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs are manufactured from yellow PE-HD plastic.

Brand:  Poeppelmann™ 70024150000

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 10093434

  • € 44.53 / Pack of 100
Estimated Shipment: 26-06-2024
to see stock.
Add to basket

Description

Description

  • With metric threads
  • Acc. with DIN EN ISO 228-1
  • Resistant to alcohol and weak acids/alkalis
  • Small to medium sizes feature a slot for screwdriver installation
  • Suitable for a variety of metric and imperial threads, as well as pipe threads
Specifications

Specifications

Screw Plug Closure
31.5 mm
Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1
M 24 x 1.5
Yellow
18 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers