missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs
Features easy-to-grip head for manual assembly. Poeppelmann™ KAPSTO™ GPN 700 Screw Plugs are manufactured from yellow PE-HD plastic.
€ 22.90 - € 117.00
Specifications
Product Type | Screw Plug Closure |
---|---|
Color | Yellow |
For Use With (Application) | Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1 |
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Closure Size | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) Outer | Height (Metric) | Closure Size | Price | Quantity & Availability | |||
10368774
|
Poeppelmann™
70008100000 |
12 mm | 12 mm | M 8 x 1 |
€ 26.60
Pack of 100 |
Estimated Shipment: 03-07-2024
Log in to see stock. |
|||
10480255
|
Poeppelmann™
70000100000 |
14 mm | 14 mm | M 10 |
€ 24.05
Pack of 100 |
Estimated Shipment: 25-06-2024
Log in to see stock. |
|||
10535674
|
Poeppelmann™
70000120000 |
16 mm | 14 mm | M 12 |
€ 24.05
Pack of 100 |
In Stock
Log in to see stock. |
|||
10003674
|
Poeppelmann™
70000140000 |
19 mm | 14 mm | M 14 |
€ 22.90
Pack of 100 |
Estimated Shipment: 18-07-2024
Log in to see stock. |
|||
10073674
|
Poeppelmann™
70016150000 |
21 mm | 16 mm | M 16 x 1.5 |
€ 30.55
Pack of 100 |
Estimated Shipment: 11-06-2024
Log in to see stock. |
|||
10606465
|
Poeppelmann™
70014150000 |
19 mm | 14 mm | M 14 x 1.5 |
€ 29.77
Pack of 100 |
Estimated Shipment: 18-06-2024
Log in to see stock. |
|||
10093434
|
Poeppelmann™
70024150000 |
31.5 mm | 18 mm | M 24 x 1.5 |
€ 44.53
Pack of 100 |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||
10378924
|
Poeppelmann™
70026150000 |
31.5 mm | 20 mm | M 26 x 1.5 |
€ 48.87
Pack of 100 |
Estimated Shipment: 05-07-2024
Log in to see stock. |
|||
10277394
|
Poeppelmann™
70045200000 |
55 mm | 25 mm | M 45 x 2 |
€ 117.00
Pack of 100 |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||
Description
- With metric threads
- Acc. with DIN EN ISO 228-1
- Resistant to alcohol and weak acids/alkalis
- Small to medium sizes feature a slot for screwdriver installation
- Suitable for a variety of metric and imperial threads, as well as pipe threads
Specifications
Screw Plug Closure | |
Yellow | |
Suitable for a variety of metric and inch size internal thread and pipe thread according to DIN EN ISO 228-1 |