Learn More
Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
€ 56.75 - € 505.00
Product Code | Brand | Model | For Use With (Equipment) | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Model | For Use With (Equipment) | Price | Quantity & Availability | ||||
10374151
|
Bandelin Electronic™
202 |
K6L | Ultrasonic Bath, RK 156 RK 156 |
€ 278.00
Each |
Estimated Shipment: 10-06-2024
Log in to see stock. |
||||
10384581
|
Bandelin Electronic™
205 |
K14B | Ultrasonic Bath, RK 514 BH, DK 514 BP |
€ 273.00
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10049290
|
Bandelin Electronic™
3025 |
K3 | Ultrasonic Bath, RK 102 CH |
€ 87.50
Each |
In Stock
Log in to see stock. |
||||
10100701
|
Bandelin Electronic™
181 |
K28C | Ultrasonic Bath, RK 1028 C/CH |
€ 364.00
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10461033
|
Bandelin Electronic™
3027 |
K5 | Ultrasonic Bath |
€ 104.00
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10717451
|
Bandelin Electronic™
138 |
K50C | Ultrasonic Bath, RK 1050 CH RK 1050 CH |
€ 505.00
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
10344301
|
Bandelin Electronic™
209 |
K08 | Ultrasonic Bath, RK/DT 31 / H |
€ 56.75
Each |
Estimated Shipment: 11-06-2024
Log in to see stock. |
||||
10733003
|
Bandelin Electronic™
3026 |
K2CL | Ultrasonic Bath, RK/DT 103 H |
€ 94.55
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
10788401
|
Bandelin Electronic™
356 |
K6 | Ultrasonic Bath, RK 106 |
€ 165.00
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10049360
|
Bandelin Electronic™
354 |
K14 | Ultrasonic Bath, RK 514/H |
€ 272.00
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
10324491
|
Bandelin Electronic™
358 |
K28 | Ultrasonic Bath, RK 1028/H / DT 1028 / H, DL 1028 H |
€ 356.00
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10458732
|
Bandelin Electronic™
359 |
K10 | Ultrasonic Bath, RK/DT 510 / 510 H / 510 H-RC, DL 510 H |
€ 258.00
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
10683401
|
Bandelin Electronic™
629 |
K6BL | Ultrasonic Bath, RK 156 BH, DK 156 BP |
€ 281.00
Each |
Estimated Shipment: 13-06-2024
Log in to see stock. |
||||
10130981
|
Bandelin Electronic™
3024 |
k1 | Ultrasonic Bath, RK 52 CH |
€ 73.30
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
||||
Description
Facilitates ultrasonic applications and simultaneously protects oscillating tank and parts to be cleaned. Bandelin Electronic™ Sonorex™ Insert Baskets for Ultrasonic Baths are manufactured from stainless steel.
- Available for a wide range of tank capacities
- Available in a range of mesh sizes
- Excellent for the cleaning of technical glassware, analysis sieves, medical instruments, metal parts, electronic components, degassing, emulsifying and sample preparation
Specifications
Basket Insert | |
Stainless Steel |