accessibility menu, dialog, popup

UserName

CAS RN 101246-66-6

CH3N(S)H3C(R)NCH3OOHN

CAS RN 101246-66-6

IUPAC Name: (3aS,8aR)-1,3a,8-trimethyl-1H,2H,3H,3aH,8H,8aH-pyrrolo[2,3-b]indol-5-yl N-phenylcarbamate
Synonyms: (3aS,8aR)-1,3a,8-trimethyl-2H,3H,8aH-pyrrolo[2,3-b]indol-5-yl N-phenylcarbamate
Molecular Weight (g/mol): 337.42
Molecular Formula: C20H23N3O2
InChi Key: PBHFNBQPZCRWQP-QUCCMNQESA-N
SMILES: CN1CC[C@]2(C)[C@H]1N(C)C1=CC=C(OC(=O)NC3=CC=CC=C3)C=C21
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results