CAS RN 101932-75-6
CAS RN 101932-75-6
IUPAC Name:
manganese(2+) bis(2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide)
Synonyms:
manganese(2+) bis(2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide)
Molecular Weight (g/mol):
297.34
Molecular Formula:
C18H26Mn
InChi Key:
FUUKHNRDHFIZLM-UHFFFAOYNA-N
SMILES:
[Mn++].CC1[C-]=C(C)C(C)=C1C.CC1[C-]=C(C)C(C)=C1C
1
–
1
of
1
results
1
Bis(tetramethylcyclopentadienyl)manganese(II), 98+%, Thermo Scientific™
CAS: 101932-75-6 Molecular Formula: C18H26Mn Molecular Weight (g/mol): 297.344 MDL Number: MFCD01862454 InChI Key: FUUKHNRDHFIZLM-UHFFFAOYSA-N Synonym: bis tetramethylcyclopentadienyl manganese,manganese 2+ bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide,manganese 2+ ion bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide PubChem CID: 74765410 IUPAC Name: manganese(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene SMILES: CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Mn+2]