Learn More
Bis(tetramethylcyclopentadienyl)manganese(II), 98+%, Thermo Scientific™
Product Code. 11461690
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. 11461690
Brand: Thermo Scientific Alfa Aesar H27027.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 101932-75-6
Molecular Formula C18H26Mn
Molecular Weight (g/mol) 297.344
MDL Number MFCD01862454
InChI Key FUUKHNRDHFIZLM-UHFFFAOYSA-N
Synonym bis tetramethylcyclopentadienyl manganese, manganese 2+ bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide, manganese 2+ ion bis 2,3,4,5-tetramethylcyclopenta-1,3-dien-1-ide
PubChem CID 74765410
IUPAC Name manganese(2+);1,2,3,5-tetramethylcyclopenta-1,3-diene
SMILES CC1[C-]=C(C(=C1C)C)C.CC1[C-]=C(C(=C1C)C)C.[Mn+2]
Specifications
Melting Point 98°C to 112°C
Quantity 5 g
Sensitivity Air and moisture sensitive
Formula Weight 297.35
Percent Purity ≥98%
Chemical Name or Material Bis(tetramethylcyclopentadienyl)manganese(II)