accessibility menu, dialog, popup

UserName

H3CONNH2

CAS RN 10201-71-5

IUPAC Name: 3-methoxypyridin-2-amine
Synonyms: 3-methoxypyridin-2-amine
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: HNAYRVKSWGSQTP-UHFFFAOYSA-N
SMILES: COC1=C(N)N=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results