Learn More
2-Amino-3-methoxypyridine, 97%, Thermo Scientific™
Product Code. 15289846 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15289846 5 g 5g
15239866 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15289846 Supplier Thermo Scientific Acros Supplier No. 453740050
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Chemical Identifiers
CAS 10201-71-5
Molecular Formula C6H8N2O
Molecular Weight (g/mol) 124.14
MDL Number MFCD07374874
InChI Key HNAYRVKSWGSQTP-UHFFFAOYSA-N
Synonym 2-amino-3-methoxypyridine, 2-pyridinamine, 3-methoxy, 3-methoxy-2-pyridylamine, 3-methoxy-pyridin-2-ylamine, 3-methoxy-2-pyridinamine, acmc-1c0wi, 3-methoxypyridine-2-amine, 3-methoxy-2-aminopyridine, 3-methoxy-pyridin-2-amine, ksc494m4p
PubChem CID 9920384
IUPAC Name 3-methoxypyridin-2-amine
SMILES COC1=C(N)N=CC=C1
Specifications
Melting Point 75°C to 77°C
Assay Percent Range 96% min. (GC)
Quantity 5 g
Formula Weight 124.14
Percent Purity 97%
Chemical Name or Material 2-Amino-3-methoxypyridine
compliance-icons
Product Identifier
  • 2-Amino-3-methoxypyridine
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.