accessibility menu, dialog, popup

UserName

CAS RN 10201-73-7

H3CONNH2

CAS RN 10201-73-7

IUPAC Name: 4-methoxypyridin-2-amine
Synonyms: 4-methoxypyridin-2-amine
Molecular Weight (g/mol): 124.14
Molecular Formula: C6H8N2O
InChi Key: QPHBCOSULYSASF-UHFFFAOYSA-N
SMILES: COC1=CC(N)=NC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results