CAS RN 102767-31-7
CAS RN 102767-31-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
(S)-N-(1-Amino-1-oxobutan-2-yl)-4-chlorobutanamide, TRC
CAS: 102767-31-7 Molecular Formula: C8 H15 Cl N2 O2 Molecular Weight (g/mol): 206.67 Synonym: Levetiracetam Related Compound A IUPAC Name: (2S)-2-(4-chlorobutanoylamino)butanamide SMILES: CC[C@H](NC(=O)CCCCl)C(=O)N
2
(S)-N-(1-Amino-1-oxobutan-2-yl)-4-chlorobutanamide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.