accessibility menu, dialog, popup

UserName

NH2ONHOOHO(Z)

CAS RN 105-61-3

IUPAC Name: (2Z)-4-(carbamoylamino)-4-oxobut-2-enoic acid
Synonyms: maleuric acid
Molecular Weight (g/mol): 158.11
Molecular Formula: C5H6N2O4
InChi Key: GWGLGTKSTGSWGQ-UPHRSURJSA-N
SMILES: NC(=O)NC(=O)\C=C/C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results