missing translation for 'onlineSavingsMsg'
Learn More

N-Carbamoylmaleamic acid, 98%, Thermo Scientific™

Product Code. 11417034
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11417034 50 g 50g
11407034 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417034 Supplier Thermo Scientific Alfa Aesar Supplier No. B21487.18
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
N-Carbamoylmaleamic acid, is used as a pharamceutical intermediate.

Solubility
Soluble in hot acetic acid, ethanol, acetone, benzene, chloroform. Practically insol in cold water, cold acetic acid, acetone, ligroin, chloroform, alc, ether.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 105-61-3
Molecular Formula C5H6N2O4
Molecular Weight (g/mol) 158.113
MDL Number MFCD00014790
InChI Key GWGLGTKSTGSWGQ-UPHRSURJSA-N
Synonym n-carbamoylmaleamic acid, maleuric acid, maleylurea, maleic acid monoureide, maleinuric acid, unii-j7d540jrqc, j7d540jrqc, 4-aminocarbonyl amino-4-oxo-2-butenoic acid, n-carbamoyl-maleamic acid, 2z-4-aminocarbonylamino-4-oxobut-2-enoic acid
PubChem CID 5356398
IUPAC Name (Z)-4-(carbamoylamino)-4-oxobut-2-enoic acid
SMILES C(=CC(=O)O)C(=O)NC(=O)N

Specifications

Melting Point 156°C to 159°C
Quantity 50 g
Beilstein 1724683
Merck Index 14,5706
Solubility Information Soluble in hot acetic acid,ethanol,acetone,benzene,chloroform. Practically insol in cold water,cold acetic acid,acetone,ligroin,chloroform,alc,ether.
Formula Weight 158.11
Percent Purity 98%
Chemical Name or Material N-Carbamoylmaleamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.