CAS RN 1092776-63-0
CAS RN 1092776-63-0
IUPAC Name:
N,N'-dicyclohexyl[({6,6-dimethyl-5H,6H-imidazo[2,1-b][1,3]thiazol-3-yl}methyl)sulfanyl]methanimidamide dihydrochloride
Synonyms:
N,N'-dicyclohexyl({6,6-dimethyl-5H-imidazo[2,1-b][1,3]thiazol-3-yl}methyl)sulfanylmethanimidamide dihydrochloride
Molecular Weight (g/mol):
479.57
Molecular Formula:
C21H36Cl2N4S2
InChi Key:
HFXJOXOIINQOEB-UHFFFAOYSA-N
SMILES:
Cl.Cl.CC1(C)CN2C(CSC(NC3CCCCC3)=NC3CCCCC3)=CSC2=N1
1
–
5
of
5
results
1
N,N'-Dicyclohexylcarbamimidothioic acid (5,6-dihydro-6,6-dimethylimidazo[2,1-b]thiazol-3-yl)methyl Ester Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N,N'-Dicyclohexylcarbamimidothioic acid (5,6-dihydro-6,6-dimethylimidazo[2,1-b]thiazol-3-yl)methyl Ester Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Tocris Bioscience™ IT1t dihydrochloride
CAS: 1092776-63-0 Molecular Formula: C21H36Cl2N4S2
4
N,N'-Dicyclohexylcarbamimidothioic acid (5,6-dihydro-6,6-dimethylimidazo[2,1-b]thiazol-3-yl)methyl Ester Dihydrochloride, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
5
IT1t dihydrochloride, MedChemExpress
MedChemExpress IT1t dihydrochloride is a potent CXCR4 antagonist; inhibits CXCL12/CXCR4 interaction with an IC50 of 2.1 nM.