CAS RN 1105193-40-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
4-(Piperazin-1-yl)-6-(pyrrolidin-1-yl)pyrimidine, TRC
CAS: 1105193-40-5 Molecular Formula: C12H19N5 Molecular Weight (g/mol): 233.31 IUPAC Name: 4-piperazin-1-yl-6-pyrrolidin-1-ylpyrimidine SMILES: C1CCN(C1)c2cc(ncn2)N3CCNCC3