accessibility menu, dialog, popup

UserName

CAS RN 112-69-6

H3CNCH3CH3

CAS RN 112-69-6

IUPAC Name: hexadecyldimethylamine
Synonyms: dimethyl palmitamine
Molecular Weight (g/mol): 269.52
Molecular Formula: C18H39N
InChi Key: NHLUVTZJQOJKCC-UHFFFAOYSA-N
SMILES: CCCCCCCCCCCCCCCCN(C)C
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results