CAS RN 1120-24-7
CAS RN 1120-24-7
IUPAC Name:
decyldimethylamine
Synonyms:
decyldimethylamine
Molecular Weight (g/mol):
185.36
Molecular Formula:
C12H27N
InChi Key:
YWWNNLPSZSEZNZ-UHFFFAOYSA-N
SMILES:
CCCCCCCCCCN(C)C
1
–
3
of
3
results
1
N,N-Dimethyldecylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N,N-Dimethyldecylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N,N-Dimethyldecylamine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.