accessibility menu, dialog, popup

UserName

ClClClOClClCl

CAS RN 116-16-5

IUPAC Name: hexachloropropan-2-one
Synonyms: hexachloroacetone
Molecular Weight (g/mol): 264.73
Molecular Formula: C3Cl6O
InChi Key: DOJXGHGHTWFZHK-UHFFFAOYSA-N
SMILES: ClC(Cl)(Cl)C(=O)C(Cl)(Cl)Cl
Molecular Weight (g/mol) 
  • (3)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (3)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)