CAS RN 1160294-93-8
CAS RN 1160294-93-8
IUPAC Name:
N-(4-bromophenyl)-[1,1'-biphenyl]-4-amine
Synonyms:
N-(4-bromophenyl)-[1,1'-biphenyl]-4-amine
Molecular Weight (g/mol):
324.22
Molecular Formula:
C18H14BrN
InChi Key:
RDSXVQDXXSPFHG-UHFFFAOYSA-N
SMILES:
BrC1=CC=C(NC2=CC=C(C=C2)C2=CC=CC=C2)C=C1
1
–
3
of
3
results
1
N-(4-Bromophenyl)-[1,1-biphenyl]-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
N-(4-Bromophenyl)-[1,1-biphenyl]-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
N-(4-Bromophenyl)-[1,1-biphenyl]-4-amine, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.