CAS RN 1163160-18-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Hippuric Acid-13C6, TRC
CAS: 1163160-18-6 Molecular Formula: C313C6H9NO3 Molecular Weight (g/mol): 185.13 Synonym: N-(Benzoyl-1,2,3,4,5,6-13C6)-glycine IUPAC Name: 2-((1,2,3,4,5,6-^{13}C_{6})cyclohexatrienecarbonylamino)acetic acid SMILES: OC(CNC([13C]1=[13C]([H])[13C]([H])=[13C]([H])[13C]([H])=[13C]1[H])=O)=O