accessibility menu, dialog, popup

UserName

CAS RN 118-93-4

IUPAC Name: 1-(2-hydroxyphenyl)ethan-1-one
Synonyms: o-acetylphenol
Molecular Weight (g/mol): 136.15
Molecular Formula: C8H8O2
InChi Key: JECYUBVRTQDVAT-UHFFFAOYSA-N
SMILES: CC(=O)C1=CC=CC=C1O
Quantity 
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
Percent Purity 
  • (1)
  • (3)
  • (2)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (2)
  • (2)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (3)