CAS RN 1189661-02-6
CAS RN 1189661-02-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Carebastine-d5, TRC
CAS: 1189661-02-6 Molecular Formula: C32H32D5NO4 Molecular Weight (g/mol): 504.67 Synonym: 4-[4-[4-(Diphenylmethoxy-d5)-1-piperidinyl]-1-oxobutyl]-α,α-dimethylbenzeneacetic Acid IUPAC Name: 2-methyl-2-[4-[4-[4-[(2,3,4,5,6-pentadeuteriophenyl)-phenylmethoxy]piperidin-1-yl]butanoyl]phenyl]propanoic acid SMILES: O=C(C1=CC=C(C(C)(C)C(O)=O)C=C1)CCCN(CC2)CCC2OC(C3=C([2H])C([2H])=C([2H])C([2H])=C3[2H])C4=CC=CC=C4