missing translation for 'onlineSavingsMsg'
Learn More

Carebastine-d5, TRC

Product Code. 30349301
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349301 1 mg 1mg
30348945 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30349301 Supplier Toronto Research Chemicals Supplier No. TRCC1834421MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1189661-02-6
Molecular Formula C32H32D5NO4
Molecular Weight (g/mol) 504.67
Synonym 4-[4-[4-(Diphenylmethoxy-d5)-1-piperidinyl]-1-oxobutyl]-α,α-dimethylbenzeneacetic Acid
IUPAC Name 2-methyl-2-[4-[4-[4-[(2,3,4,5,6-pentadeuteriophenyl)-phenylmethoxy]piperidin-1-yl]butanoyl]phenyl]propanoic acid
SMILES O=C(C1=CC=C(C(C)(C)C(O)=O)C=C1)CCCN(CC2)CCC2OC(C3=C([2H])C([2H])=C([2H])C([2H])=C3[2H])C4=CC=CC=C4

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C32H37NO4/c1-32(2,31(35)36)27-17-15-24(16-18-27)29(34)14-9-21-33-22-19-28(20-23-33)37-30(25-10-5-3-6-11-25)26-12-7-4-8-13-26/h3-8,10-13,15-18,28,30H,9,14,19-23H2,1-2H3,(H,35,36)/i3D,5D,6D,10D,11D
Chemical Name or Material Carebastine-d5
Título del producto
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.