accessibility menu, dialog, popup

UserName

CAS RN 12026-05-0

OHRb

CAS RN 12026-05-0

IUPAC Name: rubidium(1+) hydrate hydroxide
Synonyms: rubidium(1+) hydrate hydroxide
Molecular Weight (g/mol): 120.49
Molecular Formula: H3O2Rb
InChi Key: CHAWVFWLJTUJNI-UHFFFAOYSA-M
SMILES: O.[OH-].[Rb+]

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results