CAS RN 12027-38-2
CAS RN 12027-38-2
IUPAC Name:
tetracontahydrate silicon dodecatungsten
Synonyms:
silicotungstic acid
Molecular Weight (g/mol):
2954.77
Molecular Formula:
H80O40SiW12
InChi Key:
XEUBSZMTETYQPE-UHFFFAOYSA-N
SMILES:
O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.O.[Si].[W].[W].[W].[W].[W].[W].[W].[W].[W].[W].[W].[W]
1
–
1
of
1
results
1
Tungstosilicic Acid Hydrate, TRC
CAS: 12027-38-2 Molecular Formula: H4[Si(W3O10)4] . xH2O Molecular Weight (g/mol): 2896.18 Synonym: Tungstosilicic Acid (H4SiW12O40) (7CI,8CI),Silicic Acid (H4SiO4), Tungsten Complex,12-Tungstosilicic Acid,12-Tungstosilicic Acid (H4W12O40Si),Dodecatungstosilicic Acid (H4SiW12O40),JM 1493,NSW 40,PM 92,Silicotungstic Acid,Tungsten Hydroxide Oxide Silicate,(W12(OH)4O32(SiO4)),Tungsten Oxide Silicide (W12O38Si),Tungstosilicic Acid (H4W12SiO40) IUPAC Name: silicic acid;trioxotungsten SMILES: O[Si](O)(O)O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O.O=[W](=O)=O