accessibility menu, dialog, popup

UserName

NH2Cl

CAS RN 1204-44-0

IUPAC Name: 4'-chloro-[1,1'-biphenyl]-2-amine
Synonyms: 4'-chloro-[1,1'-biphenyl]-2-amine
Molecular Weight (g/mol): 203.67
Molecular Formula: C12H10ClN
InChi Key: JPBWZIPCMDZOPM-UHFFFAOYSA-N
SMILES: NC1=CC=CC=C1C1=CC=C(Cl)C=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)