CAS RN 120401-32-3
CAS RN 120401-32-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
3
of
3
results
1
6-Chloro-7-deazapurine-Alpha-D-riboside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
2
6-Chloro-7-deazapurine-Alpha-D-riboside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
6-Chloro-7-deazapurine-Alpha-D-riboside, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.