accessibility menu, dialog, popup

UserName

CAS RN 12056-07-4

SeSeSeInIn

CAS RN 12056-07-4

IUPAC Name: diindium(3+) triselandiide
Synonyms: indium(III) selenide
Molecular Weight (g/mol): 466.55
Molecular Formula: In2Se3
InChi Key: AKUCEXGLFUSJCD-UHFFFAOYSA-N
SMILES: [Se--].[Se--].[Se--].[In+3].[In+3]
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results