accessibility menu, dialog, popup

UserName

CAS RN 12070-08-5

TiC

CAS RN 12070-08-5

IUPAC Name: methane titanium
Synonyms: titanium carbide
Molecular Weight (g/mol): 63.91
Molecular Formula: CH4Ti
InChi Key: TXKRDMUDKYVBLB-UHFFFAOYSA-N
SMILES: C.[Ti]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (6)
  • (6)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)