CAS RN 1211578-68-5
IUPAC Name:
2-[3-(pentafluoro-λ⁶-sulfanyl)phenyl]acetic acid
Synonyms:
[3-(pentafluoro-λ⁶-sulfanyl)phenyl]acetic acid
Molecular Weight (g/mol):
262.19
Molecular Formula:
C8H7F5O2S
InChi Key:
MUXKXZMWGCIFAW-UHFFFAOYSA-N
SMILES:
OC(=O)CC1=CC(=CC=C1)S(F)(F)(F)(F)F
1
–
1
of
1
results
1
3-(Pentafluorothio)phenylacetic acid, 97%, Thermo Scientific™
CAS: 1211578-68-5 Molecular Formula: C8H7F5O2S Molecular Weight (g/mol): 262.194 MDL Number: MFCD16652414 InChI Key: MUXKXZMWGCIFAW-UHFFFAOYSA-N Synonym: 3-pentafluorosulfur phenylacetic acid,3-pentafluoro-??-sulfanyl phenyl acetic acid,3-pentafluorothio phenylacetic acid PubChem CID: 66523455 IUPAC Name: 2-[3-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetic acid SMILES: C1=CC(=CC(=C1)S(F)(F)(F)(F)F)CC(=O)O