missing translation for 'onlineSavingsMsg'
Learn More

3-(Pentafluorothio)phenylacetic acid, 97%, Thermo Scientific™

Product Code. 15417247
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15417247 250 mg 250mg
15407247 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15417247 Supplier Thermo Scientific Alfa Aesar Supplier No. H34302.MD
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1211578-68-5
Molecular Formula C8H7F5O2S
Molecular Weight (g/mol) 262.194
MDL Number MFCD16652414
InChI Key MUXKXZMWGCIFAW-UHFFFAOYSA-N
Synonym 3-pentafluorosulfur phenylacetic acid, 3-pentafluoro-??-sulfanyl phenyl acetic acid, 3-pentafluorothio phenylacetic acid
PubChem CID 66523455
IUPAC Name 2-[3-(pentafluoro-$l^{6}-sulfanyl)phenyl]acetic acid
SMILES C1=CC(=CC(=C1)S(F)(F)(F)(F)F)CC(=O)O

Specifications

Quantity 250 mg
Formula Weight 262.19
Percent Purity 97%
Chemical Name or Material 3-(Pentafluorothio)phenylacetic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.