accessibility menu, dialog, popup

UserName

OOH3CH3CRh

CAS RN 12245-39-5

IUPAC Name: 4-hydroxypent-3-en-2-one cycloocta-1,5-diene rhodium
Synonyms: 1,5-cyclooctadiene 4-hydroxypent-3-en-2-one rhodium
Molecular Weight (g/mol): 311.21
Molecular Formula: C13H20O2Rh
InChi Key: BUYVJWVYKPKZEX-UHFFFAOYSA-N
SMILES: [Rh].CC(O)=CC(C)=O.C1CC=CCCC=C1
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results